Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095056.2 | XP_014950542.2 | 5 - 6 | 1249 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |