Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNV1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012183958.3 | XP_012039348.2 | 1 - 2 | 624 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Ovis aries |