Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027975792.1 | XP_027831593.1 | 6 - 7 | 905 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |