Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015095779.2 | XP_014951265.2 | 1 - 2 | 126 | 112 | |
XM_027979144.1 | XP_027834945.1 | 1 - 2 | 126 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |