Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010354.4 | XP_004010403.1 | 1 - 2 | 73 | 86 | |
XM_012181018.3 | XP_012036408.1 | 1 - 2 | 64 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |