Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAKMIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960276.1 | XP_027816077.1 | 7 - 8 | 1186 | 96 | |
XM_027960278.1 | XP_027816079.1 | 7 - 8 | 1186 | 96 | ||
XM_027960280.1 | XP_027816081.1 | 7 - 8 | 1186 | 96 | ||
XM_027960282.1 | XP_027816083.1 | 7 - 8 | 1186 | 96 |