Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012526.4 | XP_004012575.1 | 3 - 4 | 695 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |