Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPERB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957272.1 | XP_027813073.1 | 16 - 17 | 1781 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |