Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027957910.1 | XP_027813711.1 | 1 - 2 | 872 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |