Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBFA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960705.1 | XP_027816506.1 | 5 - 6 | 601 | 85 | |
XM_027960706.1 | XP_027816507.1 | 4 - 5 | 416 | 85 |