Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100555.3 | XP_011955945.2 | 5 - 6 | 947 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |