Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015093090.2 | XP_014948576.2 | 3 - 4 | 1698 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |