Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101119638 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027969510.1 | XP_027825311.1 | 4 - 5 | 617 | 86 |