Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCCIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004020261.3 | XP_004020310.1 | 5 - 6 | 597 | 54 |
Gene Symbol: LOC101108057 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973235.1 | XP_027829036.1 | 1 | 580 | 54 |