Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959102.1 | XP_027814903.1 | 10 - 11 | 1420 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |