Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAXE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002621.3 | XP_004002670.2 | 6 - 7 | 909 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |