Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012097051.2 | XP_011952441.2 | 10 - 11 | 1269 | 137 | |
XM_012097052.2 | XP_011952442.2 | 10 - 11 | 999 | 137 | ||
XM_027980109.1 | XP_027835910.1 | 10 - 11 | 1062 | 137 | ||
XM_027980110.1 | XP_027835911.1 | 9 - 10 | 1401 | 137 | ||
XM_027980111.1 | XP_027835912.1 | 9 - 10 | 1401 | 137 | ||
XM_027980112.1 | XP_027835913.1 | 9 - 10 | 1401 | 137 | ||
XM_027980113.1 | XP_027835914.1 | 9 - 10 | 1401 | 137 | ||
XM_027980114.1 | XP_027835915.1 | 7 - 8 | 570 | 137 | ||
XM_027980115.1 | XP_027835916.1 | 9 - 10 | 1401 | 137 |
Gene Symbol: LOC114108742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979957.1 | XP_027835758.1 | 4 - 5 | 330 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |