Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012097051.2 | XP_011952441.2 | 6 - 7 | 844 | 90 | |
XM_012097052.2 | XP_011952442.2 | 6 - 7 | 574 | 90 | ||
XM_027980109.1 | XP_027835910.1 | 6 - 7 | 637 | 90 | ||
XM_027980110.1 | XP_027835911.1 | 5 - 6 | 976 | 90 | ||
XM_027980111.1 | XP_027835912.1 | 5 - 6 | 976 | 90 | ||
XM_027980112.1 | XP_027835913.1 | 5 - 6 | 976 | 90 | ||
XM_027980113.1 | XP_027835914.1 | 5 - 6 | 976 | 90 | ||
XM_027980114.1 | XP_027835915.1 | 3 - 4 | 145 | 90 | ||
XM_027980115.1 | XP_027835916.1 | 5 - 6 | 976 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Ovis aries |