Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOT1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004021775.4 | XP_004021824.1 | 6 - 7 | 1302 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |