Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015094347.2 | XP_014949833.2 | 4 - 5 | 367 | 65 | |
XM_015094348.2 | XP_014949834.2 | 4 - 5 | 367 | 65 | ||
XM_015094349.2 | XP_014949835.2 | 4 - 5 | 375 | 65 | ||
XM_015094350.2 | XP_014949836.2 | 4 - 5 | 367 | 65 | ||
XM_027966746.1 | XP_027822547.1 | 4 - 5 | 367 | 65 | ||
XM_027966747.1 | XP_027822548.1 | 4 - 5 | 367 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |