Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004007210.3 | XP_004007259.2 | 1 - 2 | 618 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Ovis aries |