Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6H4orf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027970859.1 | XP_027826660.1 | 6 - 7 | 1369 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |