Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012104935.3 | XP_011960325.2 | 5 - 6 | 1275 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |