Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGER4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101313.2 | XP_014956799.1 | 2 - 3 | 1459 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Ovis aries |