Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966584.1 | XP_027822385.1 | 5 - 6 | 1641 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |