Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015101913.2 | XP_014957399.1 | 8 - 9 | 1025 | 71 | |
XM_015101915.2 | XP_014957401.1 | 8 - 9 | 936 | 71 |