Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAF3IP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011198.4 | XP_004011247.1 | 3 - 4 | 1801 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |