Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012098890.3 | XP_011954280.1 | 5 - 6 | 2308 | 74 | |
XM_012098892.3 | XP_011954282.1 | 5 - 6 | 2246 | 74 | ||
XM_027957305.1 | XP_027813106.1 | 3 - 4 | 1339 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |