Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004001958.4 | XP_004002007.1 | 1 - 2 | 264 | 74 | |
XM_012138615.3 | XP_011994005.1 | 2 - 3 | 1382 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |