Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002385.4 | XP_004002434.2 | 5 - 6 | 1352 | 109 | |
XM_012179538.3 | XP_012034928.1 | 5 - 6 | 674 | 109 | ||
XM_027973642.1 | XP_027829443.1 | 5 - 6 | 1051 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |