Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004004497.4 | XP_004004546.1 | 2 - 3 | 397 | 135 | |
XM_012130961.3 | XP_011986351.1 | 2 - 3 | 646 | 135 | ||
XM_012130979.3 | XP_011986369.1 | 2 - 3 | 508 | 135 | ||
XM_027964710.1 | XP_027820511.1 | 2 - 3 | 1051 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |