Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012779.4 | XP_004012828.2 | 2 - 3 | 693 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |