Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCWPW2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027958145.1 | XP_027813946.1 | 6 - 7 | 1733 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |