Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027979950.1 | XP_027835751.1 | 4 - 5 | 741 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |