Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003046.4 | XP_004003095.2 | 2 - 3 | 421 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |