Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011650.3 | XP_004011699.2 | 3 - 4 | 459 | 78 | |
XM_027972913.1 | XP_027828714.1 | 3 - 4 | 336 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |