Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101108263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012100639.3 | XP_011956029.1 | 2 - 3 | 525 | 75 |