Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3RF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027966794.1 | XP_027822595.1 | 5 - 6 | 2303 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |