Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004010486.4 | XP_004010535.1 | 4 - 5 | 416 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |