Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015334.4 | XP_004015383.1 | 7 - 8 | 993 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |