Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006200.4 | XP_004006249.1 | 4 - 5 | 737 | 78 | |
XM_027967152.1 | XP_027822953.1 | 4 - 5 | 737 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Ovis aries |