Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008077.3 | XP_004008126.1 | 1 - 2 | 140 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |