Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004008077.3 | XP_004008126.1 | 9 - 10 | 984 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Ovis aries |