Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004016356.3 | XP_004016405.1 | 2 - 3 | 185 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |