Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974180.1 | XP_027829981.1 | 10 - 11 | 2127 | 77 | |
XM_027974181.1 | XP_027829982.1 | 10 - 11 | 2175 | 77 | ||
XM_027974182.1 | XP_027829983.1 | 9 - 10 | 2297 | 77 | ||
XM_027974183.1 | XP_027829984.1 | 10 - 11 | 2079 | 77 | ||
XM_027974184.1 | XP_027829985.1 | 10 - 11 | 2129 | 77 | ||
XM_027974186.1 | XP_027829987.1 | 11 - 12 | 1798 | 77 | ||
XM_027974187.1 | XP_027829988.1 | 11 - 12 | 1800 | 77 |