Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM20C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027961840.1 | XP_027817641.1 | 8 - 9 | 1601 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Ovis aries |