Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959867.1 | XP_027815668.1 | 6 - 7 | 1383 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |