Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRRM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012190493.3 | XP_012045883.2 | 1 | 3818 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Ovis aries |