Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027963423.1 | XP_027819224.1 | 4 - 5 | 1371 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |