Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015096596.2 | XP_014952082.2 | 2 - 3 | 378 | 81 | |
XM_015096597.2 | XP_014952083.2 | 2 - 3 | 443 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |